Learn More
Invitrogen™ Beclin 1 Recombinant Rabbit Monoclonal Antibody (24GB4550)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557347
 View more versions of this product
Product Code. 30274982
  • 564.00 € / 100µL
Description
Beclin 1 Recombinant Monoclonal Antibody for Western Blot
Beclin 1 plays a central role in autophagy. It acts as a core subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate. Different PI3K complex forms are believed to play a role in multiple membrane trafficking pathways - PI3KC3-C1 is involved in the initation of autophagosomes and PI3KC3-C2 is involved in the maturation of autophagosomes and endocytosis. Beclin 1 also plays a role in antiviral host defense.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Beclin 1
Recombinant Monoclonal
Unconjugated
BECN1
4921513J16Rik; 5430417M23Rik; APG6; Atg6; ATG6 autophagy related 6 homolog; Bcl-2-interacting protein beclin; beclin 1; beclin 1 (coiled-coil, moesin-like BCL2-interacting protein); beclin 1 (coiled-coil, myosin-like BCL2 interacting protein); beclin 1 (coiled-coil, myosin-like BCL2-interacting protein); beclin 1, autophagy related; beclin1; beclin-1; Beclin-1; beclin-1; Beclin-1-C 35 kDa; Beclin-1-C 37 kDa; becn1; Coiled-coil myosin-like BCL2-interacting protein; coiled-coil, myosin-like BCL2-interacting protein; GT197; hypothetical protein MGC73097; myosin-like BCL2 interacting protein; Protein GT197; testis secretory sperm-binding protein Li 215e; VPS30; zgc:73097
Rabbit
Affinity chromatography
RUO
114558, 56208, 8678
-20°C
Liquid
Flow Cytometry, Western Blot
24GB4550
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O88597, Q14457, Q91XJ1
BECN1
Recombinant protein of human Beclin 1 (aa 1-200).
100 μL
Primary
Human, Mouse, Rat