missing translation for 'onlineSavingsMsg'
Learn More

Behenic acid, tech. 85%

Product Code. 11485253
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485253 250 g 250g
11475253 1000 g 1000g
11495253 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485253 Supplier Thermo Scientific Alfa Aesar Supplier No. A12850.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 112-85-6 | C22H44O2 | 340.592 g/mol

Behenic acid is used to give hair conditioners and moisturizers their smoothing properties. It is also used to investigate the phase behavior of long-chain acids in supercritical propane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Behenic acid is used to give hair conditioners and moisturizers their smoothing properties. It is also used to investigate the phase behavior of long-chain acids in supercritical propane.

Solubility
Soluble in DMF (∼3 mg/ml), hot methanol, water (0.15 mg/ml at 25°C), chloroform, and ethanol (2.18 mg/ml at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, reducing agents and bases. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-85-6
Molecular Formula C22H44O2
Molecular Weight (g/mol) 340.592
MDL Number MFCD00002807
InChI Key UKMSUNONTOPOIO-UHFFFAOYSA-N
Synonym behenic acid, 1-docosanoic acid, n-docosanoic acid, hydrofol acid 560, docosoic acid, hydrofol 2022-55, glycon b-70, behensaeure, docosanoicacid, docosansaeure
PubChem CID 8215
ChEBI CHEBI:28941
IUPAC Name docosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 74°C to 78°C
Boiling Point 306°C (60 mmHg)
Quantity 250 g
Beilstein 1792887
Merck Index 14,1023
Solubility Information Soluble in DMF (∽3mg/ml),hot methanol,water (0.15mg/ml at 25°C),chloroform,and ethanol (2.18mg/ml at 25°C).
Formula Weight 340.6
Percent Purity 85%
Grade Technical
Chemical Name or Material Behenic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.