accessibility menu, dialog, popup

UserName

H3COHO

CAS RN 112-85-6

IUPAC Name: docosanoic acid
Synonyms: behenic acid zinc salt behenic acid heneicosansaeure hydrofol acid 560 behenic acid (substance) behenic acid, silver (+1) salt edenor C 22-85R potassium salt behenic acid aliphatic carboxylic acid, c22:0 c22:0 aliphatic carboxylic acid
Molecular Weight (g/mol): 340.59
Molecular Formula: C22H44O2
InChi Key: UKMSUNONTOPOIO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (1)
  • (5)
Percent Purity 
  • (5)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (5)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)