Learn More
Invitrogen™ Bemarituzumab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558313
 View more versions of this product
Product Code. 30283482
  • 477.00 € / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Bemarituzumab is a subtype-selective humanized monoclonal antibody against FGFR2b. The FIGH study aims to evaluate the efficacy of its combined chemotherapy for untreated advanced gastric cancer or gastroesophageal junction cancer.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bemarituzumab
Recombinant Monoclonal
Unconjugated
FPA114-A; FPA-144
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM Histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized