missing translation for 'onlineSavingsMsg'
Learn More

Bemcentinib, MedChemExpress

Product Code. 16476328
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476328 10 mg 10mg
16456646 100 mg 100mg
16412447 50 mg 50mg
16438917 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476328 Supplier Medchem Express Supplier No. HY15150
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bemcentinib (R428) is a potent and selective inhibitor of Axl with an IC50 of 14 nM.

Bemcentinib (R428) is a potent and selective inhibitor of Axl with an IC50 of 14 nM.

Specifications

Chemical Name or Material Bemcentinib
Physical Form Solid
CAS 1037624-75-1
Color Pink
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C30H34N8
For Use With (Application) Cancer-Kinase/protease
Synonym R428 BGB324
Solubility Information DMSO : 25 mg/mL (49.34 mM; ultrasonic and warming and heat to 60°C)
SMILES NC1=NC(NC2=CC(CC[C@@H](N3CCCC3)CC4)=C4C=C2)=NN1C(N=N5)=CC6=C5C7=CC=CC=C7CCC6
Molecular Weight (g/mol) 506.64
Formula Weight 506.64
Percent Purity 95.02%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.