missing translation for 'onlineSavingsMsg'
Learn More

Bemetizide, TRC

Product Code. 30009298
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009298 50 mg 50mg
30004392 100 mg 100mg
30007976 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009298 Supplier Toronto Research Chemicals Supplier No. TRCB13151050MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1824-52-8
Molecular Formula C15 H16 Cl N3 O4 S2
Molecular Weight (g/mol) 401.89
Synonym 6-Chloro-3,4-dihydro-3-(1-phenylethyl)-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-Dioxide, 2H-1,2,4-Benzothiadiazine-7-sulfonamide, 6-chloro-3,4-dihydro-3-(alpha-methylbenzyl)-, 1,1-dioxide (6CI,7CI,8CI), Bemeticide, Bemetizide, Dehydrosanol, Diu 60, Su 7078
IUPAC Name 6-chloro-1,1-dioxo-3-(1-phenylethyl)-3,4-dihydro-2H-1λ^{6},2,4-benzothiadiazine-7-sulfonamide
SMILES CC(C1Nc2cc(Cl)c(cc2S(=O)(=O)N1)S(=O)(=O)N)c3ccccc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H16ClN3O4S2/c1-9(10-5-3-2-4-6-10)15-18-12-7-11(16)13(24(17,20)21)8-14(12)25(22,23)19-15/h2-9,15,18-19H,1H3,(H2,17,20,21)
Formula Weight 401.0271
Percent Purity >95%
Chemical Name or Material Bemetizide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.