missing translation for 'onlineSavingsMsg'
Learn More

Benazepril Hydrochloride, TRC

Product Code. 30135043
Change view
Click to view available options
Quantity:
25 mg
50 mg
500 mg
Unit Size:
25mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135043 50 mg 50mg
30134946 500 mg 500mg
17903025 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135043 Supplier Toronto Research Chemicals Supplier No. TRCB11975050MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86541-74-4
Synonym Benazepril Hydrochloride∣(3S)-3-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid monohydrochloride∣[S-(R*,R*)]-3-[[1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid monohydrochloride∣Benazepril hydrochloride∣CGS 14824A∣CGS 14824A HCl∣Lotensin∣Lotension

Specifications

Format Neat
Molecular Weight (g/mol) 460.95
Product Type API
Quantity 50 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.