accessibility menu, dialog, popup

UserName

ClCH3OO(S)HN(S)NHOOOH

CAS RN 86541-74-4

IUPAC Name: hydrogen 2-[(3S)-3-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl]acetic acid chloride
Synonyms: hydrogen benazepril chloride
Molecular Weight (g/mol): 460.96
Molecular Formula: C24H29ClN2O5
InChi Key: VPSRQEHTHIMDQM-FKLPMGAJSA-N
SMILES: [H+].[Cl-].CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@H]1CCC2=CC=CC=C2N(CC(O)=O)C1=O
Grade 
  • (1)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)
Color 
  • (2)
Industry Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)