missing translation for 'onlineSavingsMsg'
Learn More

Benazolin-ethyl ester 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30313538
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313538 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313538 Supplier Dr. Ehrenstorfer Supplier No. DREV10450500AL100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benazolin-ethyl ester
CAS 25059-80-7
Analyte or Component Names Benazolin-ethyl ester
Shipping Condition +20°C
Molecular Formula C11 H10 Cl N O3 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C11H10ClNO3S/c1-2-16-9(14)6-13-10-7(12)4-3-5-8(10)17-11(13)15/h3-5H,2,6H2,1H3
SMILES CCOC(=O)CN1C(=O)Sc2cccc(Cl)c12
IUPAC Name ethyl 2-(4-chloro-2-oxo-1,3-benzothiazol-3-yl)acetate
Solvent or Matrix Acetonitrile
Industry Type Food Testing
Molecular Weight (g/mol) 271.72
Quantity 5 mL
Formula Weight 271.007 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.