Learn More
Descripción
Gibco DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM/F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of DMEM/F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
BenchStable DMEM/F-12 has been engineered for storage at room temperature while supporting cellular proliferation and maximum cell densities comparable to conventional DMEM/F12 when supplemented with ≥10% fetal bovine serum (FBS).
This DMEM/F-12 is modified as follows:
• Optimized for room temperature stability
• With GlutaMAX, Phenol Red
• Without HEPEPS
Using BenchStable DMEM/F-12
DMEM/F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with F-12's wide variety of components. DMEM/F-12 uses a sodium bicarbonate buffer system and therefore requires a 5–10% CO2 environment to maintain physiological pH. DMEM/F-12 with GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. BenchStable DMEM/F-12 contains no proteins, lipids, or growth factors and so requires supplementation with ≥10% fetal bovine serum (FBS).
Especificaciones
Especificaciones
| Cell Line | MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts |
| Classification | Animal Origin-free |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | DMEM (Dulbecco's Modified Eagle Medium)/F-12 |
| Serum Level | Standard Serum Supplementation |
| Sterility | Sterile-filtered |
| With Additives | High Glucose, GlutaMAX, Phenol Red, Sodium Pyruvate |
| Without Additives | No HEPES |
| Manufacturing Quality | cGMP-compliant under the ISO 9001: 2015 standard |
| Mostrar más |
For Research Use Only. Not for use in diagnostic procedures.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.