missing translation for 'onlineSavingsMsg'
Learn More

Benfotiamine, MedChemExpress

Product Code. 16461669
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461669 1 g 1g
16469817 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461669 Supplier Medchem Express Supplier No. HY17374
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Benfotiamine (S-Benzoylthiamine O-monophosphate) is an analog of vitamin B1 with higher absorption and bioavailability than vitamin B1, and is commonly used as a food supplement for diabetic complications. Benfotiamine exhibits direct antioxidative capacity and prevents induction of DNA damage.

Benfotiamine (S-Benzoylthiamine O-monophosphate) is an analog of vitamin B1 with higher absorption and bioavailability than vitamin B1, and is commonly used as a food supplement for diabetic complications. Benfotiamine exhibits direct antioxidative capacity and prevents induction of DNA damage.

Specifications

Chemical Name or Material Benfotiamine
CAS 22457-89-2
Color White
Purity Grade Notes Research
Molecular Formula C19H23N4O6PS
Quantity 1 g
Synonym S-Benzoylthiamine O-monophosphate
Solubility Information DMSO : ≥ 50 mg/mL (107.19 mM) ∣H2O : 0.67 mg/mL (1.44 mM; Need ultrasonic)
SMILES CC1=NC=C(CN(/C(C)=C(CCOP(O)(O)=O)/SC(C2=CC=CC=C2)=O)C=O)C(N)=N1
Molecular Weight (g/mol) 466.45
Formula Weight 466.45
Percent Purity 99.26%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.