missing translation for 'onlineSavingsMsg'
Learn More

Benoxaprofen, TRC

Product Code. 30132844
Change view
Click to view available options
Quantity:
1 mg
10 mg
250 mg
Unit Size:
10mg
1mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132844 250 mg 250mg
30008427 10 mg 10mg
30003311 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132844 Supplier Toronto Research Chemicals Supplier No. TRCB165300250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51234-28-7
Molecular Formula C16 H12 Cl N O3
Molecular Weight (g/mol) 301.72
Synonym (RS)-Benoxaprofen, (±)-2-(4-Chlorophenyl)-α-methyl-5-benzoxazoleacetic acid, (±)-Benoxaprofen, 2-(4-Chlorophenyl)-α-methyl-5-benzoxazoleacetic acid, 2-[2-(4-Chlorophenyl)benzoxazol-5-yl]-propanoic acid, 299582Q1, Benoxaprofen, Compound 90459, Coxigon, LRCL 3794, Lilly 90459, NSC 299582, Opren, Oraflex, Uniprofen, dl-Benoxaprofen, 5-Benzoxazoleacetic acid, 2-(4-chlorophenyl)-α-methyl-
IUPAC Name 2-[2-(4-chlorophenyl)-1,3-benzoxazol-5-yl]propanoic acid
SMILES CC(C(=O)O)c1ccc2oc(nc2c1)c3ccc(Cl)cc3

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C16H12ClNO3/c1-9(16(19)20)11-4-7-14-13(8-11)18-15(21-14)10-2-5-12(17)6-3-10/h2-9H,1H3,(H,19,20)
Formula Weight 301.0506
Chemical Name or Material Benoxaprofen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.