missing translation for 'onlineSavingsMsg'
Learn More

Bentazon, TRC

Product Code. 17930715
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17930715 100 mg 100mg
17911775 1 g 1g
17923655 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17930715 Supplier Toronto Research Chemicals Supplier No. TRCB120580100MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25057-89-0
Molecular Formula C10 H12 N2 O3 S
Molecular Weight (g/mol) 240.28
Synonym 1H-2,1,3-Benzothiadiazin-4(3H)-one, 3-isopropyl-, 2,2-dioxide (8CI), 3,4-Dihydro-3-isopropyl-1H-2,1,3-benzothiadiazin-4-one 2,2-dioxide, 3,4-Dihydro-3-isopropyl-4-oxo-1H-2,1,3-benzothiadiazine 2,2-dioxide, 3-Isopropyl-1H-2,1,3-benzothiadiazin-4(3H)-one 2,2-dioxide, 3-Isopropyl-1H-2,1,3-benzothiadiazin-4(3H)-one S,S-dioxide, 3-Isopropyl-1H-2,1,3-benzothiadiazine-4(3H)-one 2,2-dioxide, 3-Isopropyl-2,1,3-benzothiadiazin-4-one 2,2-dioxide, 3-Isopropyl-2,1,3-benzothiadiazine-4-one 2,2-dioxide, 3-Isopropyl-2,1,3-benzothiadiazinon-(4)-2,2-dioxide, 3-Isopropyl-2,1,3-benzothiodiazin-4-one 2,2-dioxide, 3-Isopropyl-4-oxo-2,1,3-benzothiadiazine 2,2-dioxide, BAS 351-07H, BAS 3510, BAS 3510H, BAS 3512H, BAS 3517H, BAS 351H, Basagran, Basagran 480, Basagran 480SL, Basamais, Bazargan, Bendioxide, Bentazon, Bentazone, Corsar, Leader
IUPAC Name 2,2-dioxo-3-propan-2-yl-1H-2λ^{6},1,3-benzothiadiazin-4-one
SMILES CC(C)N1C(=O)c2ccccc2NS1(=O)=O

Specifications

Analyte or Component Names Bentazone
Shipping Condition Room Temperature
InChI Formula InChI=1S/C10H12N2O3S/c1-7(2)12-10(13)8-5-3-4-6-9(8)11-16(12,14)15/h3-7,11H,1-2H3
Solution Type Neat
Industry Type Forensics
Quantity 100 mg
Formula Weight 240.0569
Chemical Name or Material Bentazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.