missing translation for 'onlineSavingsMsg'
Learn More

Bentazone, Dr. Ehrenstorfer

Product Code. 30319615
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319615 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319615 Supplier Dr. Ehrenstorfer Supplier No. DREC10510000100MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bentazone
CAS 25057-89-0
Analyte or Component Names Bentazone
Shipping Condition +20°C
Molecular Formula C10 H12 N2 O3 S
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C10H12N2O3S/c1-7(2)12-10(13)8-5-3-4-6-9(8)11-16(12,14)15/h3-7,11H,1-2H3
SMILES CC(C)N1C(=O)c2ccccc2NS1(=O)=O
IUPAC Name 2,2-dioxo-3-propan-2-yl-1H-2λ^{6},1,3-benzothiadiazin-4-one
Molecular Weight (g/mol) 240.28
Quantity 100 mg
Formula Weight 240.0569 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.