missing translation for 'onlineSavingsMsg'
Learn More

Benthiavalicarb Isopropyl, TRC

Product Code. 17970955
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970955 100 mg 100mg
17903455 25 mg 25mg
17941975 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17970955 Supplier Toronto Research Chemicals Supplier No. TRCB170000100MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177406-68-7
Molecular Formula C18 H24 F N3 O3 S
Molecular Weight (g/mol) 381.46
Synonym Carbamic acid, [(1S)-1-[[[(1R)-1-(6-fluoro-2-benzothiazolyl)ethyl]amino]carbonyl]-2-methylpropyl]-, 1-methylethyl ester (9CI), Carbamic acid, [1-[[[1-(6-fluoro-2-benzothiazolyl)ethyl]amino]carbonyl]-2-methylpropyl]-, 1-methylethyl ester, [S-(R*,S*)]-, 1-Methylethyl [(1S)-1-[[[(1R)-1-(6-fluoro-2-benzothiazolyl)ethyl]amino]carbonyl]-2-methylpropyl]carbamate, Bensaijunan, Benthiavalicarb isopropyl
IUPAC Name propan-2-yl N-[(2S)-1-[[(1R)-1-(6-fluoro-1,3-benzothiazol-2-yl)ethyl]amino]-3-methyl-1-oxobutan-2-yl]carbamate
SMILES CC(C)OC(=O)N[C@@H](C(C)C)C(=O)N[C@H](C)c1nc2ccc(F)cc2s1

Specifications

Analyte or Component Names Benthiavalicarb-isopropyl
Shipping Condition Room Temperature
InChI Formula InChI=1S/C18H24FN3O3S/c1-9(2)15(22-18(24)25-10(3)4)16(23)20-11(5)17-21-13-7-6-12(19)8-14(13)26-17/h6-11,15H,1-5H3,(H,20,23)(H,22,24)/t11-,15+/m1/s1
Solution Type Neat
Industry Type Environmental Testing
Quantity 100 mg
Formula Weight 381.1522
Percent Purity >95%
Chemical Name or Material Benthiavalicarb-isopropyl
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.