missing translation for 'onlineSavingsMsg'
Learn More

Benzaldehyde dimethyl acetal, 98%

Product Code. 10666622 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
missing translation for 'unitSize'
1 L
100mL
2.5 L
250mL
5mL
This item is not returnable. View return policy

Product Code. 10666622

missing translation for 'mfr': Thermo Scientific Acros 209791000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1125-88-8 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1125-88-8
MDL Number MFCD00008491
InChI Key HEVMDQBCAHEHDY-UHFFFAOYSA-N
Synonym benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca
PubChem CID 62375
IUPAC Name dimethoxymethylbenzene
SMILES COC(C1=CC=CC=C1)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.0100g/mL
Boiling Point 87.0°C to 89.0°C (18.0 mmHg)
Flash Point 69°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H12O2
Linear Formula C6H5CH(OCH3)2
Refractive Index 1.4920 to 1.4940
Quantity 100 mL
Beilstein 07,209
Specific Gravity 1.01
Solubility Information Solubility in water: decomposes
Molecular Weight (g/mol) 152.19
Formula Weight 152.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Benzaldehyde dimethyl acetal, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Benzaldehyde dimethyl acetal
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.