missing translation for 'onlineSavingsMsg'
Learn More

Benzaldehyde dimethyl acetal, 98%

Product Code. 15119233 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2.5 L
250mL
5mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15119233 2.5 L Glass bottle 2.5 L
10781481 5 mL Glass bottle 5mL
10666622 100 mL Glass bottle 100mL
10676622 250 mL Glass bottle 250mL
10135693 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15119233 Supplier Thermo Scientific Acros Supplier No. 209790025
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1125-88-8 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1125-88-8
MDL Number MFCD00008491
InChI Key HEVMDQBCAHEHDY-UHFFFAOYSA-N
Synonym benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca
PubChem CID 62375
IUPAC Name dimethoxymethylbenzene
SMILES COC(C1=CC=CC=C1)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.0100g/mL
Boiling Point 87.0°C to 89.0°C (18.0 mmHg)
Flash Point 69°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H12O2
Linear Formula C6H5CH(OCH3)2
Refractive Index 1.4920 to 1.4940
Quantity 2.5 L
Beilstein 07,209
Specific Gravity 1.01
Solubility Information Solubility in water: decomposes
Molecular Weight (g/mol) 152.19
Formula Weight 152.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Benzaldehyde dimethyl acetal, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Benzaldehyde dimethyl acetal
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.