missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution

Product Code. 10144780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10144780 1 L Glass bottle 1 L
10734034 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10144780 Supplier Thermo Scientific Acros Supplier No. 263820010
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 8001-54-5 | C9H13ClN | 170.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-54-5 , 7732-18-5
Molecular Formula C9H13ClN
Molecular Weight (g/mol) 170.66
MDL Number MFCD00145757
Synonym Alkyl-benzyl-dimethylammonium chloride

Specifications

Color Colorless to Yellow
Boiling Point >100.0°C
CAS Min % 49.0, 48.0
CAS Max % 52.0, 51.0
Concentration 49 to 52% (based on M.W.= 352.5)
Density 0.9800g/mL
Packaging Glass bottle
Specific Gravity 0.98
Viscosity 130 mPa.s (20°C)
Linear Formula C6H5CH2N(CH3)2RClR=C8H17toC18H37
pH 6 to 9 (10% aq. soln.)
Quantity 1 L
Flash Point 105°C
Merck Index 15, 1061
Physical Form Solution
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone, slightly soluble in benzene, almost insoluble in ether
Chemical Name or Material Benzalkonium chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzalkonium chloride, 50 wt% aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.