missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium Chloride, TRC

Product Code. 30138748
Change view
Click to view available options
Quantity:
2.5 g
10 g
25 g
Unit Size:
10g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138748 25 g 25g
30040816 10 g 10g
18020750 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138748 Supplier Toronto Research Chemicals Supplier No. TRCB18355025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-54-5
Formula Weight 227.1441
InChI Formula InChI=1S/C21H38N.ClH/c1-4-5-6-7-8-9-10-11-12-16-19-22(2,3)20-21-17-14-13-15-18-21;/h13-15,17-18H,4-12,16,19-20H2,1-3H3;1H/q+1;/p-1
IUPAC Name benzyl-dodecyl-dimethylazanium;chloride
Molecular Formula C11 H18 N (C2H4)n . Cl
Molecular Weight (g/mol) 227.77
SMILES C[N+](CCCCCCCCCC)(C)CC1=CC=CC=C1.Cl
Synonym Benzalkonium Chloride
Chemical Name or Material Benzalkonium chloride

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.