missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium Chloride, TRC

Product Code. 17932975
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17932975 250 mg 250mg
17962675 500 mg 500mg
17924805 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17932975 Supplier Toronto Research Chemicals Supplier No. TRCB183553250MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63449-41-2
Molecular Formula C21 H38 N . Cl
Molecular Weight (g/mol) 339.986
Synonym Benzyldimethyldecylammonium chloride, Benzenemethanaminium, N-decyl-N,N-dimethyl-, chloride, Benzenemethanaminium, N-decyl-N,N-dimethyl-, chloride (1:1), Benzyldimethyl(mixed alkyl)ammonium chloride
IUPAC Name benzyl-dodecyl-dimethylazanium;chloride
SMILES [Cl-].CCCCCCCCCCCC[N+](C)(C)Cc1ccccc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C21H38N.ClH/c1-4-5-6-7-8-9-10-11-12-16-19-22(2,3)20-21-17-14-13-15-18-21;/h13-15,17-18H,4-12,16,19-20H2,1-3H3;1H/q+1;/p-1
Formula Weight 339.269
Percent Purity >95%
Chemical Name or Material Benzyldimethyldecylammonium chloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.