missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonamide, 98+%

Product Code. 10167253 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10167253 100 g Plastic bottle 100g
10153290 5 g Glass bottle 5g
10605202 500 g Plastic bottle 500g
10712105 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10167253 Supplier Thermo Scientific Acros Supplier No. 148491000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-10-2 | C6H7NO2S | 157.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-10-2
Molecular Formula C6H7NO2S
Molecular Weight (g/mol) 157.19
MDL Number MFCD00007930
InChI Key KHBQMWCZKVMBLN-UHFFFAOYSA-N
Synonym benzenesulphonamide, benzosulfonamide, benzolsulfonamide, phenyl sulfonamide, benzene sulfonamide, phenylsulfonamide, m and b 7973, chembl27601, benzolsulfonamid, benzensulfonamide
PubChem CID 7370
IUPAC Name benzenesulfonamide
SMILES C1=CC=C(C=C1)S(=O)(=O)N

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 151°C to 153°C
Color White
Flash Point 250°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5SO2NH2
Quantity 100 g
Beilstein 11, 39
Solubility Information Solubility in water: 4.3g/L (16°C)
Formula Weight 157.19
Percent Purity 98+%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Benzenesulfonamide, 98+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.