missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonyl hydrazide, 98%

Product Code. 11496846
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496846 100 g 100g
11486846 25 g 25g
11406856 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496846 Supplier Thermo Scientific Alfa Aesar Supplier No. B23971.22
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80-17-1 | C6H8N2O2S | 172.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-17-1
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00007583,MFCD08064450
InChI Key VJRITMATACIYAF-UHFFFAOYSA-N
Synonym benzenesulfonyl hydrazide, porofor, benzenesulfonic acid, hydrazide, porofor bsh, benzenesulfohydrazide, phenylsulfonylhydrazine, genitron bsh, celogen bsh, benzenesulfonylhydrazine, phenylsulfohydrazide
PubChem CID 65723
IUPAC Name benzenesulfonohydrazide
SMILES NNS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 101°C to 110°C (decomposition)
Quantity 100 g
UN Number UN3226
Beilstein 640079
Merck Index 14,7598
Formula Weight 172.21
Percent Purity 98%
Chemical Name or Material Benzenesulfonyl hydrazide

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.