missing translation for 'onlineSavingsMsg'
Learn More

Benzethonium chloride, 97%

Product Code. 10619113 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10619113 2.5 kg Plastic drum 2.5kg
10366020 100 g Plastic bottle 100g
10497470 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10619113 Supplier Thermo Scientific Acros Supplier No. 105380025
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

Benzethonium chloride, 97%, C27H42ClNO2, CAS Number-121-54-0, benzethoniumchloride, benzethonium chloride, disilyn, hyamine, kylacol, phemeride, phemerol chloride, phemithyn, quatrachlor, solamin, 2.5kg, 96% min. (on dry substance) (Argentometry), 100.0, 96.0, CHEBI:31264, White, 204-479-9, 01,433 | CAS: 121-54-0 | C27H42ClNO2 | 448.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-54-0
Molecular Formula C27H42ClNO2
Molecular Weight (g/mol) 448.08
MDL Number MFCD00011742
InChI Key UREZNYTWGJKWBI-UHFFFAOYSA-M
Synonym benzethonium chloride, quatrachlor, hyamine, phemeride, phemerol chloride, benzethoniumchloride, phemithyn, disilyn, kylacol, solamin
PubChem CID 8478
ChEBI CHEBI:31264
IUPAC Name benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
SMILES CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 158.0°C to 163.0°C
Color White
Loss on Drying 5% max. (105°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (on dry substance) (Argentometry)
Packaging Plastic drum
Quantity 2.5 kg
Fieser 01,433
Merck Index 15, 1076
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,acetone and chloroform
Formula Weight 448.08
Percent Purity 97%
Physical Form Amorphous or Crystalline Powder
Chemical Name or Material Benzethonium chloride, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzethonium chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 C
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.