missing translation for 'onlineSavingsMsg'
Learn More

Benzethonium Chloride, 98%, MP Biomedicals

Product Code. 11473006 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473006 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11473006 Supplier MP Biomedicals Supplier No. 0522166110
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 121-54-0
Molecular Formula C27H42ClNO2
Quantity 10 g
Synonym benzethonium chloride, quatrachlor, hyamine, phemeride, phemerol chloride, benzethoniumchloride, phemithyn, disilyn, kylacol, solamin
InChI Key UREZNYTWGJKWBI-UHFFFAOYSA-M
SMILES CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]
IUPAC Name benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
Molecular Weight (g/mol) 448.088
PubChem CID 8478
ChEBI CHEBI:31264
Formula Weight 466.11
Chemical Name or Material Benzethonium Chloride, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.