missing translation for 'onlineSavingsMsg'
Learn More

Benzethonium chloride, BAKER ANALYZED™, J.T. Baker™

Product Code. 15558404 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15558404 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15558404 Supplier JT Baker Supplier No. 7170.1000
  • ONLINE EXCLUSIVE PRICE
    42.60 €
    35.20 € / 1 L
    Save 7.40 €
    17% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Laboratory Reagent

  • Titer: 0.0039 - 0.0041 mol/L

Specifications

Packaging Glass bottle
CAS 121-54-0
Molecular Formula C27H42ClNO2
Concentration 0.004 mol/L
MDL Number MFCD00011742
Synonym Hyamine™ 1622
InChI Key UREZNYTWGJKWBI-UHFFFAOYSA-M
SMILES [Cl-].CC(C)(C)CC(C)(C)C1=CC=C(OCCOCC[N+](C)(C)CC2=CC=CC=C2)C=C1
IUPAC Name benzyldimethyl(2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethyl)azanium chloride
Molecular Weight (g/mol) 448.09
Quantity 1 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.