missing translation for 'onlineSavingsMsg'
Learn More

Benzhydrol, 99%

Product Code. 11405333
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405333 250 g 250g
11495323 50 g 50g
11485323 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405333 Supplier Thermo Scientific Alfa Aesar Supplier No. A12884.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 91-01-0 | C13H12O | 184.238 g/mol

Benzhydrol is widely used as intermediates in pharmaceuticals (including antihistamines), agrochemicals, perfumes and other organic compounds. It is used as a fixative in the perfume industry. It is involved in polymerization reaction as a terminating group. It is used as precursor to prepare modafinil, benztropine and diphehydramine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzhydrol is widely used as intermediates in pharmaceuticals (including antihistamines), agrochemicals, perfumes and other organic compounds. It is used as a fixative in the perfume industry. It is involved in polymerization reaction as a terminating group. It is used as precursor to prepare modafinil, benztropine and diphehydramine.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, acids, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-01-0
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.238
MDL Number MFCD00004488
InChI Key QILSFLSDHQAZET-UHFFFAOYSA-N
Synonym benzhydrol, diphenylcarbinol, benzohydrol, benzhydryl alcohol, hydroxydiphenylmethane, diphenylmethyl alcohol, diphenyl carbinol, alpha-phenylbenzenemethanol, diphenyl-methanol, benzenemethanol, .alpha.-phenyl
PubChem CID 7037
IUPAC Name diphenylmethanol
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)O

Specifications

Melting Point 64°C to 68°C
Boiling Point 297°C to 298°C
Flash Point 160°C (320°F)
Odor Odorless
Quantity 250 g
Beilstein 1424379
Merck Index 14,1090
Solubility Information Slightly soluble in water.
Formula Weight 184.24
Percent Purity 99%
Chemical Name or Material Benzhydrol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.