missing translation for 'onlineSavingsMsg'
Learn More

Benzhydryl isothiocyanate, ≥90%, Thermo Scientific™

Product Code. 10651113
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10651113 10 g 10g
10163954 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10651113 Supplier Thermo Scientific Maybridge Supplier No. S03388EA
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 3550-21-8
Molecular Formula C14H11NS
Molecular Weight (g/mol) 225.31
MDL Number MFCD00046815
InChI Key WDOSFTZMBFYTED-UHFFFAOYSA-N
Synonym benzhydryl isothiocyanate, isothiocyanatodiphenylmethane, benzhydrylisothiocyanate, isothiocyanato phenyl methyl benzene, diphenylmethylisothiocyanate, benzene, 1,1'-isothiocyanatomethylene bis, diphenylmethanisothiocyanate, acmc-20alq6, benzhydryl-isothiocyanate, benzhydryl isothio-cyanate
PubChem CID 520742
IUPAC Name [isothiocyanato(phenyl)methyl]benzene
SMILES S=C=NC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Linear Formula C6H5CH(NCS)C6H5
Quantity 10 g
Formula Weight 225.31
Percent Purity 90%
Chemical Name or Material Benzhydryl isothiocyanate
compliance-icons
Product Identifier
  • Benzhydryl isothiocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.