missing translation for 'onlineSavingsMsg'
Learn More

Benzo[a]pyrene, 96%

Product Code. 11337148
Change view
Click to view available options
Quantity:
100 mg
0.5 g
Unit Size:
0.5g
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337148 0.5 g 0.5g
11347148 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337148 Supplier Thermo Scientific Alfa Aesar Supplier No. 015856.02
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Extensively used in cancer research | CAS: 50-32-8 | C20H12 | 252.32 g/mol

Extensively used in cancer research and for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. It is also a multipurpose intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-32-8
Molecular Formula C20H12
Molecular Weight (g/mol) 252.32
MDL Number MFCD00003602
InChI Key FMMWHPNWAFZXNH-UHFFFAOYSA-N
Synonym benzo a pyrene, 3,4-benzopyrene, benzo pqr tetraphene, 3,4-benzpyrene, benzpyrene, benzo def chrysene, 6,7-benzopyrene, benz a pyrene, 3,4-benz a pyrene
PubChem CID 2336
ChEBI CHEBI:29865
IUPAC Name benzo[a]pyrene
SMILES C1=CC=C2C(=C1)C=C1C=CC3=CC=CC4=CC=C2C1=C34

Specifications

Melting Point 177°C to 180°C
Boiling Point 495°C
Quantity 0.5 g
UN Number UN3077
Merck Index 14,1103
Solubility Information Soluble in benzene, toluene, and xylene; Sparingly soluble in alcohol, methanol; Insoluble in water
Formula Weight 252.3
Percent Purity 96%
Physical Form Crystalline
Chemical Name or Material Benzo[a]pyrene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.