missing translation for 'onlineSavingsMsg'
Learn More

Benzo[a]pyrene Diol Epoxide (>90%), TRC

Product Code. 30004537
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004537 2.5 mg 2.5mg
30008670 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004537 Supplier Toronto Research Chemicals Supplier No. TRCB2875502.5MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58917-67-2
Molecular Formula C20 H14 O3
Molecular Weight (g/mol) 302.3234
Synonym (7R,8S,8aS,9aR)-rel-7,8,8a,9a-Tetrahydrobenzo[10,11]chryseno[3,4-b]oxirene-7,8-diol, (7α,8β,8aα,9aα)-7,8,8a,9aTetrahydrobenzo[10,11]chryseno[3,4-b]oxirene-7,8-diol, (±)-7α,8β-Dihydroxy-9β,10β-epoxy-7,8,9,10-tetrahydrobenzo[a]pyrene, (±)-7β,8α-Dihydroxy-9α,10α-epoxy-7,8,9,10-tetrahydrobenzo[a]pyrene, (±)-anti-BPDE, anti-Benzo[a]pyrene-7,8-diol-9,10-epoxide, trans-7,8-Dihydroxy-anti-9,10-epoxy-7,8,9,10-tetrahydrobenzo[a]pyrene
IUPAC Name (3R,5S,6S,7R)-4-oxahexacyclo[11.6.2.02,8.03,5.010,20.017,21]henicosa-1(20),2(8),9,11,13(21),14,16,18-octaene-6,7-diol
SMILES O[C@@H]1[C@@H]2O[C@@H]2c3c(cc4ccc5cccc6ccc3c4c56)[C@H]1O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C20H14O3/c21-17-13-8-11-5-4-9-2-1-3-10-6-7-12(15(11)14(9)10)16(13)19-20(23-19)18(17)22/h1-8,17-22H/t17-,18+,19-,20+/m1/s1
Formula Weight 302.0943
Chemical Name or Material Benzo[a]pyrene Diol Epoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.