missing translation for 'onlineSavingsMsg'
Learn More

Benzo[a]pyrene, MedChemExpress

Product Code. 16444118
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16444118 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16444118 Supplier Medchem Express Supplier No. HY107377
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Benzo[a]pyrene shows lung carcinogenicity in animal models, and it is frequently used in chemoprevention studies.

Benzo[a]pyrene shows lung carcinogenicity in animal models, and it is frequently used in chemoprevention studies.

Specifications

Chemical Name or Material Benzo[a]pyrene
CAS 50-32-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H12
Quantity 10 mg
Synonym 3,4-Benzopyrene
Solubility Information DMSO : 50 mg/mL (198.17 mM; Need ultrasonic and warming)
SMILES C12=C(C=C3)C=C(C=CC=C4)C4=C1C=CC5=CC=CC3=C25
Molecular Weight (g/mol) 252.31
Formula Weight 252.31
Percent Purity 98.75%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.