missing translation for 'onlineSavingsMsg'
Learn More

Benzo(a)pyrene, SPEX CertiPrep™

Product Code. 12909434 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12909434 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12909434 Supplier SPEX Certiprep Supplier No. S430
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Benzo(a)pyrene50-32-81000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging 2mL amber prescored glass ampule
CAS 50-32-8, 75-09-2
CAS Min % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molecular Formula C20H12
MDL Number MFCD00003602
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key FMMWHPNWAFZXNH-UHFFFAOYSA-N
Type Certified Reference Material
SMILES C1=CC=C2C(=C1)C=C1C=CC3=CC=CC4=CC=C2C1=C34
IUPAC Name pentacyclo[10.6.2.02,7.09,19.016,20]icosa-1(18),2,4,6,8,10,12,14,16,19-decaene
Solvent or Matrix Methylene Chloride
Molecular Weight (g/mol) 252.32
PubChem CID 2336
Quantity 1 mL
ChEBI CHEBI:29865
Chemical Name or Material Benzo(a)pyrene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzo(a)pyrene
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Acute toxicity Category 4
Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: dichloromethane, benzo[a]pyrene

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.