missing translation for 'onlineSavingsMsg'
Learn More

Benzo(g,h,i)perylene, SPEX CertiPrep™

Product Code. 16369577 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16369577 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16369577 Supplier SPEX Certiprep Supplier No. S445

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beinhaltet

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Benzo(g,h,i)perylene191-24-21000μg/mL

Zertifizierungen

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Spezifikation

Packaging 2mL Amber Prescored Glass Ampule
CAS 191-24-2, 75-09-2
CAS Min % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molecular Formula C22H12
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key GYFAGKUZYNFMBN-UHFFFAOYSA-N
SMILES C1=CC2=C3C(=C1)C4=CC=CC5=C4C6=C(C=C5)C=CC(=C36)C=C2
Solvent or Matrix Methylene Chloride
Molecular Weight (g/mol) 276.338
PubChem CID 9117
Quantity 1 mL
ChEBI CHEBI:34568
Chemical Name or Material Benzo(g,h,i)perylene
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Benzo(g,h,i)perylene
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Flammable liquid Category 2
Gefahrenhinweis
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H351-Suspected of causing cancer.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Ergänzende Informationen
  • MIXTURE LIST-Contain: dichloromethane

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.