missing translation for 'onlineSavingsMsg'
Learn More

Benzo[ghi]perylene, 98+%

Product Code. 10105643 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10105643 5 mg Glass bottle 5mg
10206523 25 mg Glass bottle 25mg
10498430 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10105643 Supplier Thermo Scientific Acros Supplier No. 105550050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 191-24-2 | C22H12 | 276.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 191-24-2
Molecular Formula C22H12
Molecular Weight (g/mol) 276.33
MDL Number MFCD00004135
InChI Key GYFAGKUZYNFMBN-UHFFFAOYSA-N
Synonym benzo ghi perylene, 1,12-benzoperylene, 1,12-benzperylene, benzo g,h,i perylene, benzo ghi pyrilene, benzo-1,12-perylene, unii-5zme2e2q9l, ccris 784
PubChem CID 9117
ChEBI CHEBI:34568
SMILES C1=CC2=C3C(=C1)C4=CC=CC5=C4C6=C(C=C5)C=CC(=C36)C=C2

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 276.0°C to 280.0°C
Color Yellow
Boiling Point >500.0°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 5 mg
Solubility Information Solubility in water: insoluble
Formula Weight 276.33
Percent Purity 98+%
Physical Form Needle-like Crystals or Crystalline Powder
Chemical Name or Material Benzo[ghi]perylene, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • Benzo[ghi]perylene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.