missing translation for 'onlineSavingsMsg'
Learn More

Benzo[ghi]perylene, TRC

Product Code. 17991975
Change view
Click to view available options
Quantity:
25 mg
100 mg
5 g
Unit Size:
100mg
25mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17991975 100 mg 100mg
17922965 5 g 5g
17915065 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17991975 Supplier Toronto Research Chemicals Supplier No. TRCB207700100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 191-24-2
Molecular Formula C22 H12
Molecular Weight (g/mol) 276.33
Synonym Benzo[ghi]perylene, 1,12-Benzoperylene, 1,12-Benzperylene, NSC 89275
IUPAC Name Benzo[g,h,i]perylene
SMILES c1cc2ccc3ccc4ccc5cccc6c(c1)c2c3c4c56

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C22H12/c1-3-13-7-9-15-11-12-16-10-8-14-4-2-6-18-17(5-1)19(13)21(15)22(16)20(14)18/h1-12H
Formula Weight 276.0939
Percent Purity >95%
Chemical Name or Material Benzo[ghi]perylene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.