missing translation for 'onlineSavingsMsg'
Learn More

Benzoic acid, 99.6%, ACS reagent

Product Code. 10005860 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
25g
2kg
500g
This item is not returnable. View return policy

Product Code. 10005860

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65-85-0 | C7H6O2 | 122.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-85-0
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
InChI Key WPYMKLBDIGXBTP-UHFFFAOYSA-N
Synonym benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv
PubChem CID 243
ChEBI CHEBI:30746
IUPAC Name benzoic acid
SMILES C1=CC=C(C=C1)C(=O)O

Specifications

CAS Min % 99.5
CAS Max % 100.0
Melting Point 121.0°C to 123.0°C
Color White
Boiling Point 249.0°C
Flash Point 121°C
Ignition Residue 0.005% max.
Infrared Spectrum Authentic
Assay Percent Range 99.6%
Packaging Plastic bottle
Linear Formula C6H5CO2H
Quantity 500 g
Beilstein 09,92
Fieser 01,49; 13,312
Merck Index 15,1093
Solubility Information Solubility in water: 3g/L (20°C). Other solubilities: soluble in alcohol,benzene,ether,chloroform,,ccl4,carbon disulfide and turpentine
Formula Weight 122.12
Percent Purity 99.6%
Grade ACS Reagent
Insoluble Matter 0.005% max. (in methanol)
Physical Form Crystalline Powder
Chemical Name or Material Benzoic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzoic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Benzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.