missing translation for 'onlineSavingsMsg'
Learn More

Benzoic acid, 99%, extra pure

Product Code. 10137603 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Unit Size:
1kg
2.5kg
25kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10137603 500 g Plastic bottle 500g
10326840 1 kg Plastic bottle 1kg
10455365 2.5 kg Plastic bottle 2.5kg
15149193 25 kg Fibre drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10137603 Supplier Thermo Scientific Acros Supplier No. 149135000
Chemical Monthly Promotion
Use promo code 28093
View all promotions
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

Benzoic acid, 99%, C7H6O2, CAS Number-65-85-0, benzenecarboxylic acid, benzeneformic acid, benzenemethanoic acid, benzoesaeure gk, benzoesaeure gv, carboxybenzene, dracylic acid, phenylcarboxylic acid, phenylformic acid, retardex, 2.5kg, (20 wt.% soln. in ethanol) clear, 09,92, 249.0 deg.C, 100.0 | CAS: 65-85-0 | C7H6O2 | 122.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-85-0
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00002398
InChI Key WPYMKLBDIGXBTP-UHFFFAOYSA-N
Synonym benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv
PubChem CID 243
ChEBI CHEBI:30746
IUPAC Name benzoic acid
SMILES C1=CC=C(C=C1)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 121.0°C to 123.0°C
Color Orange to White
Boiling Point 249.0°C
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C6H5CO2H
Quantity 500 g
Beilstein 09,92
Fieser 01,49; 13,312
Merck Index 15,1093
Solubility Information Solubility in water: 3g/L (20°C). Other solubilities: soluble in alcohol,benzene,ether,chloroform,,ccl4,carbon disulfide and turpentine
Formula Weight 122.12
Clarity (20 wt.% soln. in ethanol) clear
Percent Purity 99%
Grade Extra Pure
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Benzoic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzoic acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.