missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Acid, Certified AR for Analysis, Fisher Chemical™

Product Code. 10650321 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10650321 1 kg 1kg
10476860 100 g 100g
10172180 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10650321 Supplier Fisher Chemical Supplier No. B/1950/60
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Benzoic Acid, Certified AR for Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-85-0
Molecular Formula C7H6O2
MDL Number 2398

Specifications

Melting Point 121°C
Color White
pH 2.5 to 3.5
Boiling Point 249°C
Packaging Plastic powder jar
Quantity 1 kg
Formula Weight 122.12g/mol
Physical Form Solid
compliance-icons compliance-icons
Product Identifier
  • Benzoic acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.