missing translation for 'onlineSavingsMsg'
Learn More

Benzoic acid, MedChemExpress

Product Code. 16419036
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16419036 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16419036 Supplier Medchem Express Supplier No. HYN0216
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Benzoic acid is an aromatic alcohol existing naturally in many plants and is a common additive to food, drinks, cosmetics and other products. It acts as preservatives through inhibiting both bacteria and fungi.

Benzoic acid is an aromatic alcohol existing naturally in many plants and is a common additive to food, drinks, cosmetics and other products. It acts as preservatives through inhibiting both bacteria and fungi.

Specifications

Chemical Name or Material Benzoic acid
Physical Form Solid
CAS 65-85-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C7H6O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (818.87 mM) ∣H2O : 7.14 mg/mL (58.47 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=CC=C1
Molecular Weight (g/mol) 122.12
Formula Weight 122.12
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.