missing translation for 'onlineSavingsMsg'
Learn More

Benzoic anhydride, 98%

Product Code. 10297493 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10297493 100 g Plastic bottle 100g
10697432 25 g Plastic bottle 25g
10204660 500 g Plastic bottle 500g
11994971 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10297493 Supplier Thermo Scientific Acros Supplier No. 105501000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 93-97-0 | C14H10O3 | 226.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 93-97-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00003073
InChI Key CHIHQLCVLOXUJW-UHFFFAOYSA-N
Synonym benzoic anhydride, benzoic acid, anhydride, benzoyl anhydride, benzoylbenzoate, benzoic acid anhydride, benzoic acid, 1,1'-anhydride, phenyl anhydride, benzoesaeureanhydrid, unii-9k7x34fov2, phenylcarbonyl benzoate
PubChem CID 7167
ChEBI CHEBI:38815
IUPAC Name benzoyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OC(=O)C2=CC=CC=C2

Especificaciones

CAS Min % 97.5
CAS Max % 100.0
Melting Point 38.0°C to 44.0°C
Color White
Density 1.1900g/mL
Boiling Point 360.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula (C6H5CO)2O
Quantity 100 g
Beilstein 09,164
Fieser 01,49; 05,23
Merck Index 15,1094
Specific Gravity 1.19
Solubility Information Solubility in water: 0,01g/L. Other solubilities: soluble in alcohol,chloroform,acetone,benzene,,ethyl acetate,toluene,xylene,ether,glacial,acetic acid,acetic anhydride,moderately soluble in petroleum ether
Formula Weight 226.23
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Benzoic anhydride, 98%
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • Benzoic anhydride
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.