missing translation for 'onlineSavingsMsg'
Learn More

Benzoin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30312701
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 30312701

Brand: Dr. Ehrenstorfer DREV10538250AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzoin
CAS 119-53-9
Analyte or Component Names Benzoin
Shipping Condition +20°C
Molecular Formula C14 H12 O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Ethanone, 2-hydroxy-1,2-diphenyl-, Benzoin (8CI), 2-Hydroxy-1,2-diphenylethanone, (RS)-Benzoin, (±)-2-Hydroxy-1,2-diphenylethanone, (±)-2-Hydroxy-2-phenylacetophenone, (±)-Benzoin, 1,2-Diphenyl-1-hydroxy-2-ethanone, 1,2-Diphenyl-2-hydroxyethanone, 1,2-Diphenyl-2-oxoethanol, 2-Hydroxy-1,2-diphenyl-1-ethanone, 2-Hydroxy-1-phenyl-2-phenylethanone, 2-Hydroxy-2-phenylacetophenone, 2-Hydroxyl-1,2-phenylethanone, 2-Phenyl-2-hydroxyacetophenone, AW 63, Benzoylphenylcarbinol, DL-Benzoin, Desyl alcohol, Esacure EB 3, F 297, GK 19, Jiqi GK 19, L 307, LC Benzoin, NSC 8082, Nisso Cure MB, S 19, S 19 (benzoin), Seikuol Z, Wy 42956, dl-Benzoin, α-Benzoylbenzenemethanol, α-Hydroxy-α-phenylacetophenone, α-Hydroxybenzyl phenyl ketone
InChI Formula InChI=1S/C14H12O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13,15H
SMILES OC(C(=O)c1ccccc1)c2ccccc2
IUPAC Name 2-hydroxy-1,2-diphenylethanone
Solvent or Matrix Acetonitrile
Industry Type Food Testing
Molecular Weight (g/mol) 212.24
Quantity 5 mL
Formula Weight 212.0837 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.