missing translation for 'onlineSavingsMsg'
Learn More

Benzoin, Extra Pure, SLR, Fisher Chemical

Product Code. 10753981 Shop All Fisher Chemical Products
Change view
Click to view available options
250g, Plastic powder jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10753981 250g, Plastic powder jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10753981 Supplier Fisher Chemical Supplier No. B/2050/50
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 119-53-9
Melting Point 133°C
Color White
Boiling Point 194°C
Packaging Plastic powder jar
Molecular Formula C14H12O2
MDL Number 4496
Quantity 250 g
Formula Weight 212.25g/mol
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.