missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone, 99%

Product Code. 11479612
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479612 5000 g 5000g
11469612 250 g 250g
11459612 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479612 Supplier Thermo Scientific Alfa Aesar Supplier No. A10739.0I
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 119-61-9 | C13H10O | 182.222 g/mol

Benzophenone is widely employed as a photo initiator in UV-curing applications such as inks, imaging and clear coatings in the printing industry. It acts as a UV blocker to prevent photo-degradation of the packaging polymers or its contents. It protects some products such as perfumes and soaps from ultraviolet light. It serves to manufacturers to package the product in clear glass or plastic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzophenone is widely employed as a photo initiator in UV-curing applications such as inks, imaging and clear coatings in the printing industry. It acts as a UV blocker to prevent photo-degradation of the packaging polymers or its contents. It protects some products such as perfumes and soaps from ultraviolet light. It serves to manufacturers to package the product in clear glass or plastic.

Solubility
Soluble in ethanol and diethyl ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-61-9
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.222
MDL Number MFCD00003076
InChI Key RWCCWEUUXYIKHB-UHFFFAOYSA-N
Synonym benzophenone, diphenyl ketone, benzoylbenzene, phenyl ketone, methanone, diphenyl, alpha-oxoditane, ketone, diphenyl, benzene, benzoyl, alpha-oxodiphenylmethane, diphenylketone
PubChem CID 3102
ChEBI CHEBI:41308
IUPAC Name diphenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 47°C to 50°C
Density 1.111
Boiling Point 305°C
Flash Point 138°C (280°F)
Assay Percent Range 99%
Odor Pleasant
Quantity 5000 g
UN Number UN3077
Beilstein 1238185
Merck Index 14,1098
Solubility Information Soluble in ethanol and diethyl ether. Insoluble in water.
Formula Weight 182.22
Percent Purity 99%
Chemical Name or Material Benzophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.