missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone imine, 97%, stabilized

Product Code. 10111152 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10111152 5 g Glass bottle 5g
10049041 25 g Glass bottle 25g
10593674 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10111152 Supplier Thermo Scientific Acros Supplier No. 368590050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1013-88-3 | C13H11N | 181.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1013-88-3 , 123-31-9
Molecular Formula C13H11N
Molecular Weight (g/mol) 181.23
MDL Number MFCD00001760
InChI Key SXZIXHOMFPUIRK-UHFFFAOYSA-N
Synonym benzophenone imine, benzophenoneimine, benzhydrylideneamine, 1,1-diphenylmethanimine, unii-ejj21na7vi, benzhydrylimine, benzophenonimine, diphenylmethanimineimine, 1,1-diphenylmethylimine, benzenemethanimine, .alpha.-phenyl
PubChem CID 136809
IUPAC Name diphenylmethanimine
SMILES C1=CC=C(C=C1)C(=N)C2=CC=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -30°C
Color Brown-Yellow to Colorless
Density 1.0800g/mL
Boiling Point 216°C (75.0 mmHg)
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2C=NH
Refractive Index 1.6170 to 1.619
Quantity 5 g
Beilstein 07, IV, 1365
Merck Index 15, 1102
Specific Gravity 1.08
Solubility Information Solubility in water: 0.3g/L (25°C). Other solubilities: better soluble in aq. solution at lower ph values,,very soluble in diethyl ether
Stabilizer approx. 0.02% w/w hydroquinone
Viscosity 33,4 mPa.s (20°C)
Formula Weight 181.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Benzophenone imine, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzophenone imine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.