missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone, TRC

Product Code. 30004708
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004708 10 g 10g
17995055 1 g 1g
17932565 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004708 Supplier Toronto Research Chemicals Supplier No. TRCB20498010G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-61-9
Molecular Formula C13 H10 O
Molecular Weight (g/mol) 182.22
Synonym Benzophenone (7CI,8CI), Diphenylmethanone, Adjutan 6016, BLS 531, BP, Benzene, benzoyl-, Benzoylbenzene, Darocur BP, Diphenyl ketone, Irgacure BP, JRCure 1020, Kayacure BP, Kayacure BP 100, Lowlite 24, NSC 8077, Phenyl ketone, Runtecure 1020, Speedcure BP, Winure BP, α-Oxodiphenylmethane, α-Oxoditane
IUPAC Name diphenylmethanone
SMILES O=C(c1ccccc1)c2ccccc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H
Formula Weight 182.0732
Percent Purity >95%
Chemical Name or Material Benzophenone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.