missing translation for 'onlineSavingsMsg'
Learn More

Benzophenonetetracarboxylic acid, MedChemExpress

Product Code. 16499616
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499616 50 mg 50mg
16464356 100 mg 100mg
16443058 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16499616 Supplier Medchem Express Supplier No. HY100511
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Benzophenonetetracarboxylic acid (3,3',4,4'-Benzophenonetetracarboxylic acid) is particularly useful in the preparation of high performance polyimides and also useful as curing agents for epoxy resins.

Benzophenonetetracarboxylic acid (3,3',4,4'-Benzophenonetetracarboxylic acid) is particularly useful in the preparation of high performance polyimides and also useful as curing agents for epoxy resins.

Specifications

Chemical Name or Material Benzophenonetetracarboxylic acid
CAS 2479-49-4
Color White
Purity Grade Notes Research
Molecular Formula C17H10O9
Quantity 50 mg
Synonym 3,3',4,4'-Benzophenonetetracarboxylic acid
Solubility Information DMSO : 50 mg/mL (139.56 mM; Need ultrasonic) ∣H2O : 10 mg/mL (27.91 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C(O)=O)C(C(O)=O)=C1)C2=CC=C(C(O)=O)C(C(O)=O)=C2
Molecular Weight (g/mol) 358.26
Formula Weight 358.26
Percent Purity 98.66%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.