missing translation for 'onlineSavingsMsg'
Learn More

Benzopyrene, TRC

Product Code. 30136364
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136364 1 g 1g
18022230 250 mg 250mg
18020050 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136364 Supplier Toronto Research Chemicals Supplier No. TRCB2058001G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-32-8
Molecular Formula C20 H12
Molecular Weight (g/mol) 252.31
Synonym Benzo[a]pyrene, 3,4-Benz[a]pyrene, 3,4-Benzopyrene, 3,4-Benzpyrene, 3,4-Benzpyrene (carcinogen), 4,5-Benzpyrene, 6,7-Benzopyrene, BaP, Benz[a]pyrene, Benzo[d,e,f]chrysene, Benzopyrene, NSC 21914
IUPAC Name benzo[a]pyrene
SMILES c1ccc2c(c1)cc3ccc4cccc5ccc2c3c45

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H12/c1-2-7-17-15(4-1)12-16-9-8-13-5-3-6-14-10-11-18(17)20(16)19(13)14/h1-12H
Formula Weight 252.0939
Percent Purity >95%
Chemical Name or Material Benzo[a]pyrene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.