missing translation for 'onlineSavingsMsg'
Learn More

Benzopyrene, TRC

Product Code. 18022230
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18022230 250 mg 250mg
30136364 1 g 1g
18020050 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18022230 Supplier Toronto Research Chemicals Supplier No. TRCB205800250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-32-8
Formula Weight 252.0939
InChI Formula InChI=1S/C20H12/c1-2-7-17-15(4-1)12-16-9-8-13-5-3-6-14-10-11-18(17)20(16)19(13)14/h1-12H
IUPAC Name benzo[a]pyrene
Molecular Formula C20 H12
Molecular Weight (g/mol) 252.31
Percent Purity >95%
SMILES c1ccc2c(c1)cc3ccc4cccc5ccc2c3c45
Synonym Benzo[a]pyrene, 3,4-Benz[a]pyrene, 3,4-Benzopyrene, 3,4-Benzpyrene, 3,4-Benzpyrene (carcinogen), 4,5-Benzpyrene, 6,7-Benzopyrene, BaP, Benz[a]pyrene, Benzo[d,e,f]chrysene, Benzopyrene, NSC 21914
Chemical Name or Material Benzo[a]pyrene
Grade HPLC
Show More Show Less

Specifications

Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.