missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl isothiocyanate, 98%

Product Code. 11327076
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11327076

Brand: Thermo Scientific Alfa Aesar L06964.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 532-55-8 | C8H5NOS | 163.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-55-8
Molecular Formula C8H5NOS
Molecular Weight (g/mol) 163.194
MDL Number MFCD00004815
InChI Key CPEKAXYCDKETEN-UHFFFAOYSA-N
Synonym benzoylthiocarbimide, benzoylisothiocyanate, benzoylisothiocyante, isothiocyanic acid benzoyl ester, unii-u8j7q32b86, n-benzoyl isothiocyanate, benzoic acid, anhydride with hncs, benzoic acid, anhydride with isothiocyanic acid, oxophenylmethanisothiocyanate, 1-benzoyl acetone
PubChem CID 68284
IUPAC Name benzoyl isothiocyanate
SMILES C1=CC=C(C=C1)C(=O)N=C=S

Specifications

Density 1.214
Boiling Point 128°C to 131°C (15 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.6375
Quantity 25 g
UN Number UN2922
Beilstein 606716
Sensitivity Moisture sensitive
Merck Index 14,1114
Formula Weight 163.2
Percent Purity 98%
Chemical Name or Material Benzoyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.