missing translation for 'onlineSavingsMsg'
Learn More

Benzoylformic acid, 97%

Product Code. 10755361 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10755361 5 g Glass bottle 5g
10396160 25 g Glass bottle 25g
10182270 100 g Glass bottle 100g
11965101 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10755361 Supplier Thermo Scientific Acros Supplier No. 105760050
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 611-73-4 | C8H6O3 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-73-4
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.13
MDL Number MFCD00002575
InChI Key FAQJJMHZNSSFSM-UHFFFAOYSA-N
Synonym benzoylformic acid, phenylglyoxylic acid, oxophenylacetic acid, benzeneglyoxylic acid, phenylgloxylic acid, 2-phenylethanoic acid, oxo phenyl acetic acid, benzoylformate, phenylglyoxalate, phenyloxoacetic acid
PubChem CID 11915
ChEBI CHEBI:18280
IUPAC Name 2-oxo-2-phenylacetic acid
SMILES OC(=O)C(=O)C1=CC=CC=C1

Specifications

Melting Point 60°C to 69°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5COCO2H
Quantity 5 g
Beilstein 10,654
Solubility Information Solubility in water: .. Other solubilities: 1g/10mL methanol
Formula Weight 150.13
Percent Purity 97%
Physical Form Adhering Crystals, Crystalline Powder or Chunks
Chemical Name or Material Benzoylformic acid
Show More Show Less
compliance-icons
Product Identifier
  • Benzoylformic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.