missing translation for 'onlineSavingsMsg'
Learn More

Benzoylurea, 97%, Thermo Scientific™

Product Code. 11347696
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347696 5 g 5g
11357696 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347696 Supplier Thermo Scientific Alfa Aesar Supplier No. L11313.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-22-2
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.16
MDL Number MFCD00025441
InChI Key HRYILSDLIGTCOP-UHFFFAOYSA-N
Synonym benzoylurea, 1-benzoylurea, n-benzoylurea, urea, benzoyl, benzoic acid, ureide, n-aminocarbonyl benzamide, benzamide, n-aminocarbonyl, benzamide, n-aminocarbonyl-9ci, benzoyl urea, benzoylcarbamide
PubChem CID 69190
ChEBI CHEBI:39416
IUPAC Name N-carbamoylbenzamide
SMILES NC(=O)NC(=O)C1=CC=CC=C1

Specifications

Melting Point 209°C to 210°C
Quantity 5 g
Beilstein 2045538
Formula Weight 164.17
Percent Purity 97%
Chemical Name or Material Benzoylurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.